Learn More
Leica Biosystems 7ml IgM Bond RTU Primary
Supplier:  Leica Biosystems PA0278U
Encompass
Antigen BackgroundIgM, together with IgD, is the major immunoglobulin expressed on the surface of B cells and normally constitutes about 1 per cent of serum immunoglobulin. IgM antibody is prominent in early immune responses to most antigens and predominates in certain antibody responses such as natural blood group antibodies.DisclaimerImmunoglobulin M is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2117
 Restricted Item