Learn More
Leica Biosystems 7ML MAST CELL TRYP BONDRTUPRIM
Supplier:  Leica Biosystems PA0019
Encompass
Mast cells contain a number of preformed chemical mediators such as histamine, chymase, carboxypeptidase and proteolytic tryptase. A substantial quantity of tryptase is reported to be found in mast cells of skin and lung and suggests this enzyme plays a major role in mast cell mediated events. In vitro studies indicate tryptase can cleave C3 to form C3a anaphylatoxin, inactivate fibrinogen as a coaguable substrate for thrombin and activate latent collagenase. Models of allergenic disease in the skin, nose and lung have each indicated elevated tryptase levels. Human mast cell tryptase has been reported to be implicated as a mediator of inflammation. Mast cell degranulation in the gut causes mucus secretion, mucosal edema, increased gut permeability and may be responsible for some of the symptoms and signs of inflammatory bowel disease.
Catalog No. NC2156099