Learn More
Leica Biosystems 7ML Napsin A (MRQ-60 Mab) 'CM'
Supplier:  Leica Biosystems PA0805
Encompass
This antibody is intended for in vitro diagnostic (IVD) use. The Cell Marque Napsin A (MRQ-6) antibody is intended for qualified laboratories to qualitatively identify by light microscopy the presence of associated antigens in sections of formalin-fixed, paraffin-embedded tissue sections using IHC test methods. Use of this antibody is indicated, subsequent to clinical differential diagnosis, as an aid in the identification of napsin A protein over-expression in various lung adenocarcinomas within the context of an antibody panel, the patient’s clinical history, and other diagnostic tests evaluated by a qualified pathologist.
Catalog No. 50-255-2036