Learn More
Leica Biosystems 7ml p53 Bond RTU Primary
Supplier:  Leica Biosystems PA0057U
Encompass
Antigen BackgroundThis monoclonal antibody recognizes both wild type and mutant forms of human p53 protein under denaturing and non-denaturing conditions. The epitope recognized by clone DO-7 can be destroyed by prolonged fixation in buffered formalin. The heat induced epitope retrieval technique may improve staining in some cases.Disclaimerp53 Protein is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2166