Learn More
Bellco Glass, Inc. 7mL Pipette, MLA, Macro, Red
SDP
Catalog No. 502330087
Encompass_Preferred
7mL Pipette, MLA, Macro, RedSingle-stroke “to deliver” operationAvailable in 10 fixed volumes ranging from 1 mL- 10 mLAlso available as 1-5 mL Adjustable model with 0.1 mL incrementsAlso available as Selectable model for set volumes: 2 mL/3 mL/4 mL/5 mL All pipettes in stock for immediate shipmentFind tip size for your MLA pipette chartCase Quantity1Primary ColorRedPrimary MaterialAluminum Alloy & Stainless SteelVolume7mLVistaLab Tip Fit SizeMacro 10Universal FitNoAccuracy±0.60%Precision0.30%
Catalog No. 50-233-0087 Supplier Bellco Glass, Inc. Supplier No. 451900086
7mL Pipette, MLA, Macro, RedSingle-stroke “to deliver” operationAvailable in 10 fixed volumes ranging from 1 mL- 10 mLAlso available as 1-5 mL Adjustable model with 0.1 mL incrementsAlso available as Selectable model for set volumes: 2 mL/3 mL/4 mL/5 mL All pipettes in stock for immediate shipmentFind tip size for your MLA pipette chartCase Quantity1Primary ColorRedPrimary MaterialAluminum Alloy & Stainless SteelVolume7mLVistaLab Tip Fit SizeMacro 10Universal FitNoAccuracy±0.60%Precision0.30%