Learn More
Sarstedt Inc 7ML SC TUBE 13X82 FBGWB/PK1000
Supplier:  Sarstedt Inc 62.550.008
Encompass_Preferred
7mL Screen Printed Screw Cap Tube 13x82mm Flat Bottom Polypropylene Graduations and Writing Block Neutral Cap
Catalog No. NC9588468