Learn More
Sarstedt Inc 7ML SCTUBE 13X82 RB GWB/CS1000
Supplier:  Sarstedt Inc 62.550.032
Encompass_Preferred
7ml Screen Printed Screw Cap Tube 13x82mm Round Bottom Polypropylene Graduations and Writing Block
Catalog No. NC9017622