Learn More
Uline Transfer Pipette - 7 mL | | 500
Catalog No. NC2949195
Encompass
Transfer fluids to sample tubes one drop at a time, Use in classrooms and research labs Non-sterile, Squeezable and flexible LDPE plastic, Disposable to prevent cross-contamination, Graduated for accurate measurement
Catalog No. NC2949195 Supplier Uline Supplier No. S24320
Transfer fluids to sample tubes one drop at a time, Use in classrooms and research labs Non-sterile, Squeezable and flexible LDPE plastic, Disposable to prevent cross-contamination, Graduated for accurate measurement