Learn More
CELLTREAT Scientific Products 7mL Transfer Pipet, Graduated to 3mL, Bulk Bag, Non-sterile, Case Quantity 500
SDP
Catalog No. 5000670030
Encompass_Preferred
Transfer Pipets are easy to manipulate with a gentle squeeze, and do not require any additional equipment
Translucent and flexible pipet body is ideal for a range of liquid handling applications
Each pipet features molded graduations with labeled increments
Sterile Transfer Pipets are conveniently individually wrapped in plastic and are suitable for transferring sterilesolutions or cell suspensions
Non-sterile bulk Transfer Pipets are ideal for regular benchtop use
Catalog No. 50-006-70030 Supplier CELLTREAT Scientific Products Supplier No. 229217
Transfer Pipets are easy to manipulate with a gentle squeeze, and do not require any additional equipment
Translucent and flexible pipet body is ideal for a range of liquid handling applications
Each pipet features molded graduations with labeled increments
Sterile Transfer Pipets are conveniently individually wrapped in plastic and are suitable for transferring sterilesolutions or cell suspensions
Non-sterile bulk Transfer Pipets are ideal for regular benchtop use