Learn More
Leica Biosystems 7ml Vimentin Bond RTU Primary
Supplier:  Leica Biosystems PA0640U
Encompass
Antigen BackgroundEukaryotic cells contain a number of types of cytoplasmic filamentous proteins, microtubule, microfilaments and intermediate-sized filaments (IF). Vimentin, a 57 kD protein that is an intermediate filament is reported to be expressed in most cells of mesenchymal origin, including fibroblasts, endothelial cells, smooth muscle, melanocytes as well as T and B lymphocytes.DisclaimerVimentin is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2059