Learn More
Ace Glass, Inc. 7MM GRADED SEALS 7MM GRADED SEALS
SDP
Catalog No. 50974592
Encompass
7mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-974-592 Supplier Ace Glass, Inc. Supplier No. 845510
7mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.