Learn More
Ace Glass, Inc. 7MM JOINT (GB)
SDP
Catalog No. 50975235
Encompass_Preferred
7mm glass o-ring seal joint tooled with a groove to take a -111 FETFE o-ring, and to provide a greaseless, vacuum-tight connection. Supplied with one o-ring, which may be used up to 230°C.
Catalog No. 50-975-235 Supplier Ace Glass, Inc. Supplier No. 764604
7mm glass o-ring seal joint tooled with a groove to take a -111 FETFE o-ring, and to provide a greaseless, vacuum-tight connection. Supplied with one o-ring, which may be used up to 230°C.