Learn More
Ace Glass, Inc. #7 Ace-Thred solid PTFE plug, 7855-707 FETFE O-ring front seal
SDP
Catalog No. 509557163
Encompass_Preferred
#7 Ace-Thred solid PTFE plug with front seal O-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -009 FETFE O-ring.
Catalog No. 50-955-7163 Supplier Ace Glass, Inc. Supplier No. 584644
#7 Ace-Thred solid PTFE plug with front seal O-ring, for sealing column ends. Permits preparation and storage of columns. Supplied with size -009 FETFE O-ring.