Learn More
Fine Science Tools Reflex Wound Clips, Stainless Steel, 7 x 3 mm, 100/pk
SDP
Catalog No. NC9400060
Encompass
Product: Reflex Wound Clips - 7mm/ Item: 7mm Clips/ Use with: 7mm Applicator/ Dimensions: 7 x 3mm/ Alloy: Stainless Steel/ Sterile: No/ Unit: 100
Catalog No. NC9400060 Supplier Fine Science Tools Supplier No. 1203207
Product: Reflex Wound Clips - 7mm/ Item: 7mm Clips/ Use with: 7mm Applicator/ Dimensions: 7 x 3mm/ Alloy: Stainless Steel/ Sterile: No/ Unit: 100