Learn More
Perkin Elmer US LLC 7-Position Standards Rack for 250 mL Bottles
Catalog No. 502117212
Encompass_Preferred
Rack can be used with Polyethylene Wide Mouth Bottles (N0777818).
Catalog No. 50-211-7212 Supplier Perkin Elmer US LLC Supplier No. N8122464
Rack can be used with Polyethylene Wide Mouth Bottles (N0777818).