Learn More
Medchemexpress LLC 7PPD-Q | 2894124-00-4 | 98.9% | 312.41 | 1 MG
SDP
Catalog No. 5000390575
Encompass_Preferred
7PPD-Q is a substituted p-phenylenediamine antioxidant derivative. It has been found to be toxic to the bacterium *V. fischeri* with an EC50 of 14.9 mg/L and is intended for research use only.
  • Acts as an antioxidant derivative
  • Exhibits toxicity to *Vibrio fischeri* bacterium
  • Suitable for research applications
  • Available as a solid in pale purple to purple color
  • Soluble in DMSO for in vitro studies
  • Can be formulated for in vivo studies with DMSO and corn oil
Catalog No. 50-003-90575 Supplier Medchemexpress LLC Supplier No. HY1678871MG
7PPD-Q is a substituted p-phenylenediamine antioxidant derivative. It has been found to be toxic to the bacterium *V. fischeri* with an EC50 of 14.9 mg/L and is intended for research use only.
  • Acts as an antioxidant derivative
  • Exhibits toxicity to *Vibrio fischeri* bacterium
  • Suitable for research applications
  • Available as a solid in pale purple to purple color
  • Soluble in DMSO for in vitro studies
  • Can be formulated for in vivo studies with DMSO and corn oil