Learn More
Medchemexpress LLC 7PPD-Q | 2894124-00-4 | 98.9% | 312.41 | 5 MG
SDP
Catalog No. 5000391141
Encompass_Preferred
7PPD-Q is a substituted p-phenylenediamine antioxidants derivative. It has been observed to be toxic to the bacterium V. fischeri (EC50 = 14.9 mg/L).
  • For research use only.
  • Not sold to patients.
Catalog No. 50-003-91141 Supplier Medchemexpress LLC Supplier No. HY1678875MG
7PPD-Q is a substituted p-phenylenediamine antioxidants derivative. It has been observed to be toxic to the bacterium V. fischeri (EC50 = 14.9 mg/L).
  • For research use only.
  • Not sold to patients.