Learn More
Medchemexpress LLC (7S)-BAY-593 | 2413068-25-2 | 99.6% | 476.53 | 100 MG
SDP
Catalog No. 5000394023
Encompass_Preferred
(7S)-BAY-593 is the S-enantiomer of BAY-593, an orally active GGTase-I inhibitor. It effectively blocks YAP1/TAZ signaling in animals and demonstrates significant antitumor activity.
  • S-enantiomer of BAY-593
  • Orally active GGTase-I inhibitor
  • Blocks YAP1/TAZ signaling
  • Exhibits antitumor activity
  • Targets YAP/TAZ-TEAD and GGTase I
  • Soluble in DMSO
Catalog No. 50-003-94023 Supplier Medchemexpress LLC Supplier No. HY161573A100MG
(7S)-BAY-593 is the S-enantiomer of BAY-593, an orally active GGTase-I inhibitor. It effectively blocks YAP1/TAZ signaling in animals and demonstrates significant antitumor activity.
  • S-enantiomer of BAY-593
  • Orally active GGTase-I inhibitor
  • Blocks YAP1/TAZ signaling
  • Exhibits antitumor activity
  • Targets YAP/TAZ-TEAD and GGTase I
  • Soluble in DMSO