Learn More
Medchemexpress LLC (7S)-BAY-593 | 2413068-25-2 | 99.61% | 476.53 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY161573A50MG
Encompass_Preferred
(7S)-BAY-593 is the S-enantiomer of BAY-593 (HY-161573). BAY-593 is an orally active GGTase-I inhibitor. BAY-593 can block YAP1/TAZ signaling in animals and has antitumor activity.
Catalog No. 50-003-95321