Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ 7X 6 Pack

Catalog No. MP0976680A2 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
1 pk (6x6.4 oz)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP0976680A2 1 pk (6x6.4 oz)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP0976680A2 Supplier MP Biomedicals™ Supplier No. 0976680A2

missing translation for 'validMainframeMsg'

7X 6 PACK, Liquid

  • 7X Ready-to-Use requires no pH adjustment or titration.
  • 7X 6 Pack are ready-to-use premeasured refills.
  • Pour one refill bottle into an empty gallon container, then completely fill with purified water. The Refill 6-Pack yields 6 gallons of 5% concentration 7X.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Lab Essentials
Form Liquid
Content And Storage Store at Room Temperature. (15°C to 30°C)
Quantity 1 Ea.
Volume (English) 6.4 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.