Learn More
Edmund Optics 7X MEAS LOUPE W/LIN SCALE
Catalog No. NC0660326
Encompass
7X Peak Measuring Loupe with linear scale. Includes removable glass reticle scale that should be placed in direct contact with object. Features smooth helicoid twist focus and achromatic lens system for color corrected flat field viewing.
Catalog No. NC0660326 Supplier Edmund Optics Supplier No. 35325
7X Peak Measuring Loupe with linear scale. Includes removable glass reticle scale that should be placed in direct contact with object. Features smooth helicoid twist focus and achromatic lens system for color corrected flat field viewing.