Learn More
Electron Microscopy Sciences 7x Measuring Magnifier Metric Scale
SDP
Catalog No. 5027059
Encompass_Preferred
Achromatic lens with a photo-printed precise scale, focusing ring, and objective. Lens: 19mm, eyepiece: 18mm, scale diameter: 26mm measuring scale: 20mm, divisions: 0.1mm, divisions: 0.005". Metric & English scale interchangeable. Size: 36mm
Catalog No. 50-270-59 Supplier Electron Microscopy Sciences Supplier No. 68150
Achromatic lens with a photo-printed precise scale, focusing ring, and objective. Lens: 19mm, eyepiece: 18mm, scale diameter: 26mm measuring scale: 20mm, divisions: 0.1mm, divisions: 0.005". Metric & English scale interchangeable. Size: 36mm