Learn More
Bellco Glass, Inc. 7X-O-Maticモ Detergent,1 Gal
SDP
Catalog No. 501945676
Encompass_Preferred
7X detergent is a liquid concentrate at pH 7.8 of anionic surfactants and special solvent for use in washing procedures for all laboratory glass ware. Its non-caustic non-acidic and indicated for hand washing procedures. Gallon: 1
Catalog No. 50-194-5676 Supplier Bellco Glass, Inc. Supplier No. 475220001
7X detergent is a liquid concentrate at pH 7.8 of anionic surfactants and special solvent for use in washing procedures for all laboratory glass ware. Its non-caustic non-acidic and indicated for hand washing procedures. Gallon: 1