Learn More
Ace Glass, Inc. 7X7MM RASCHIG RINGS 7X7MM RASCHIG RINGS
SDP
Catalog No. 50979622
Encompass_Preferred
7 x 7mm raschig rings are fabricated from borosilicate glass and have an approximate wall weight of 1.0/mm.
Catalog No. 50-979-622 Supplier Ace Glass, Inc. Supplier No. 803308
7 x 7mm raschig rings are fabricated from borosilicate glass and have an approximate wall weight of 1.0/mm.