Learn More
Sklar Instruments Single Use, Econo-Sterile, Rochester-Pean Forceps,Sklar 8-1/2 in., Blunt, Curved, Horizontally Serrated, Stainless Steel, Disposable. CS of 25.
SDP
Catalog No. 501094263 Shop All Sklar Instruments Products
Encompass_Preferred
Single Use, Econo-Sterile, Rochester-Pean Forceps,Sklar, 8-1/2 in., Blunt,Curved, Horizontally Serrated, Stainless Steel, Disposable. CS of 25. Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.
Catalog No. 50-109-4263 Supplier Sklar Instruments Supplier No. 962548
Single Use, Econo-Sterile, Rochester-Pean Forceps,Sklar, 8-1/2 in., Blunt,Curved, Horizontally Serrated, Stainless Steel, Disposable. CS of 25. Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.