Learn More
Sklar Instruments Forceps, Birkett Hemostat Forceps, Sklar 8-1/2 in., Straight, Serrated, Finger Rings, Stainless Steel, EA.
SDP
Catalog No. 501097946 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Birkett Hemostat Forceps, Sklar, 8-1/2 in., Straight, Serrated, Finger Rings, Stainless Steel, EA. Sklar Birkett Hemostatic Forceps are slender, ratcheted, finger ring forceps used for clamping vessels and controlling blood flow. They are similar to the Moynihan hemostat, with fully serrated jaws and commonly used in procedures in the abdominal area; however, the Birkett hemostat has slightly larger jaws and is often slightly longer. This product is straight with an overall length of 8-1/2 inches.
Catalog No. 50-109-7946 Supplier Sklar Instruments Supplier No. 173212
Forceps, Birkett Hemostat Forceps, Sklar, 8-1/2 in., Straight, Serrated, Finger Rings, Stainless Steel, EA. Sklar Birkett Hemostatic Forceps are slender, ratcheted, finger ring forceps used for clamping vessels and controlling blood flow. They are similar to the Moynihan hemostat, with fully serrated jaws and commonly used in procedures in the abdominal area; however, the Birkett hemostat has slightly larger jaws and is often slightly longer. This product is straight with an overall length of 8-1/2 inches.