Learn More
Chemglass Life Sciences Stopcock, Straight Bore, Glass Shell only for PTFE Plug, 8mm & 10mm Bore, 24/40 Barrel Size, 16mm OD Sidearms
SDP
Catalog No. CG42608
Encompass_Preferred
Stopcock, Straight Bore, Glass Shell only for PTFE Plug, 8mm & 10mm Bore, 24/40 Barrel Size, 16mm OD Sidearms. Glass portion only of CG-424 stopcock. Accepts 1:5 taper PTFE plugs.
Catalog No. CG42608 Supplier Chemglass Life Sciences Supplier No. CG42608
Stopcock, Straight Bore, Glass Shell only for PTFE Plug, 8mm & 10mm Bore, 24/40 Barrel Size, 16mm OD Sidearms. Glass portion only of CG-424 stopcock. Accepts 1:5 taper PTFE plugs.