Learn More
Medchemexpress LLC 8,11,14,17-eicosatetraenoic acid ethyl ester | 123940-93-2 | 98.0% | 332.52 g·mol⁻¹ | C22H36O2 | 500UG
SDP
Catalog No. 5000237651
Encompass_Preferred
Omega-3 arachidonic acid ethyl ester is the ethyl-ester derivative of arachidonic acid provided as a high-purity solution in ethanol for biochemical and lipid research applications. The material is characterized by CAS 123940-93-2, molecular formula C22H36O2, and molecular weight 332.52 g·mol⁻¹, and is supplied with recommended storage at -20°C.
Catalog No. 50-002-37651 Supplier Medchemexpress LLC Supplier No. HY125139500UG
Omega-3 arachidonic acid ethyl ester is the ethyl-ester derivative of arachidonic acid provided as a high-purity solution in ethanol for biochemical and lipid research applications. The material is characterized by CAS 123940-93-2, molecular formula C22H36O2, and molecular weight 332.52 g·mol⁻¹, and is supplied with recommended storage at -20°C.