Learn More
Medchemexpress LLC 8,11,14-Eicosatrienamide, N-(2-hydroxyethyl)-, (8Z,11Z,14Z)- | 150314-34-4 | MFCD00674172 | ≥99.0% | 349.55 g·mol⁻1 | C22H39NO2 | 5 MG
SDP
Catalog No. 5000262993
Encompass_Preferred
Dihomo-γ-linolenoyl ethanolamide is an endogenous N-acylethanolamine that acts as a cannabinoid (CB) receptor agonist. It binds human recombinant CB1 and CB2 receptors with reported Ki values of 857 nM and 598 nM, respectively. The compound is provided as a high-purity research reagent for biochemical and pharmacology studies.
  • Acts as a cannabinoid receptor agonist (CB1 and CB2).
  • Reported Ki values: CB1 857 nM, CB2 598 nM.
  • High purity (≥99.0% by HPLC).
  • Molecular formula C22H39NO2; molecular weight 349.55 g·mol⁻1.
  • Available in small-scale research quantities, e.g., 5 mg.
  • Suitable for endocannabinoid signaling and receptor pharmacology research.
Catalog No. 50-002-62993 Supplier Medchemexpress LLC Supplier No. HY1130705MG
Dihomo-γ-linolenoyl ethanolamide is an endogenous N-acylethanolamine that acts as a cannabinoid (CB) receptor agonist. It binds human recombinant CB1 and CB2 receptors with reported Ki values of 857 nM and 598 nM, respectively. The compound is provided as a high-purity research reagent for biochemical and pharmacology studies.
  • Acts as a cannabinoid receptor agonist (CB1 and CB2).
  • Reported Ki values: CB1 857 nM, CB2 598 nM.
  • High purity (≥99.0% by HPLC).
  • Molecular formula C22H39NO2; molecular weight 349.55 g·mol⁻1.
  • Available in small-scale research quantities, e.g., 5 mg.
  • Suitable for endocannabinoid signaling and receptor pharmacology research.