Learn More
Cayman Chemical 8 12-Iso-IPF2a-VI 10ug
SDP
Catalog No. 502972428
Encompass_Preferred
The most abundant isoprostane product formed during lipid peroxidation; detected after CCl4-induced oxidative damage to hepatic tissue and has been found elevated in the urine, blood, and CSF of patients with Alzheimer’s disease
Catalog No. 50-297-2428 Supplier Cayman Chemical Supplier No. 1631010 UG
The most abundant isoprostane product formed during lipid peroxidation; detected after CCl4-induced oxidative damage to hepatic tissue and has been found elevated in the urine, blood, and CSF of patients with Alzheimer’s disease