Learn More
Cayman Chemical 8 12isoiPF2aVI1 5lactone
SDP
Supplier:  Cayman Chemical 1031250 UG
Encompass_Preferred
A racemic mixture of the lactone form of the free acid, 8,12-iso- iPF2α-VI; the free acid is the most abundant F2-iP2 regioisomer measured in the urine of rats treated to induce lipid peroxidation; used as a biomarker for oxidative stress
Catalog No. 50-288-7425