Learn More
Cayman Chemical 8 12isoiPF2aVId11
SDP
Catalog No. 502890975
Encompass_Preferred
An internal standard for the quantification of 8,12-iso-iPF2α-VI by GC- or LC-MS.
Catalog No. 50-289-0975 Supplier Cayman Chemical Supplier No. 1000687850 UG
An internal standard for the quantification of 8,12-iso-iPF2α-VI by GC- or LC-MS.