Learn More
Cayman Chemical 8 12isoiPF2aVId11
SDP
Supplier:  Cayman Chemical 1000687825 UG
Encompass_Preferred
An internal standard for the quantification of 8,12-iso-iPF2α-VI by GC- or LC-MS.
Catalog No. 50-288-9927