Learn More
Avanti Polar Lipids 8(14)-DEHYDROCHOLESTEROL 1MG
Supplier:  Avanti Polar Lipids 700076P1MG
Encompass
8(14)-dehydrocholesterol, >95% 1mg CAS#177962-82-2 C27H44O FW:384.638
Catalog No. NC2198160