Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8,14-methano-2h,14h-1-benzopyrano[7,8-d]benzodioxocin-3,5,11,13,15-pentol, 2,8-bis(3,4-di | 103883-03-0 | 99.2% | 576.50 | C30H24O12 | 10 MG
SDP

Catalog No. 5000289901
Encompass_Preferred

Procyanidin A1 is a procyanidin dimer provided as a research/analytical standard for biochemical and cell biology studies, with activity in PKC-related and inflammation/immunology assays.

  • High purity (HPLC) 99.2%.
  • Molecular formula C30H24O12; molecular weight 576.50.
  • CAS number 103883-03-0.
  • Supplied as a 10 MG research vial.
  • Store solid at 4°C and protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Intended for in vitro and analytical applications; batch COA available for assay and appearance.

Catalog No. 50-002-89901 Supplier Medchemexpress LLC Supplier No. HYN234410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Procyanidin A1 is a procyanidin dimer provided as a research/analytical standard for biochemical and cell biology studies, with activity in PKC-related and inflammation/immunology assays.

  • High purity (HPLC) 99.2%.
  • Molecular formula C30H24O12; molecular weight 576.50.
  • CAS number 103883-03-0.
  • Supplied as a 10 MG research vial.
  • Store solid at 4°C and protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
  • Intended for in vitro and analytical applications; batch COA available for assay and appearance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.