Learn More
Bio Rad Laboratories 8–16% Criterion TGX Precast Midi Protein Gel, 26 well, 15 uL
Catalog No. NC1313021
Encompass
Pkg of 1, 8–16% precast polyacrylamide gel, 13.3 × 8.7 cm (W × L), for use with Criterion and Criterion Dodeca electrophoresis cells.
Catalog No. NC1313021 Supplier Bio Rad Laboratories Supplier No. 5671105
Pkg of 1, 8–16% precast polyacrylamide gel, 13.3 × 8.7 cm (W × L), for use with Criterion and Criterion Dodeca electrophoresis cells.