Learn More
Medchemexpress LLC 8-(2-((2-pentylcyclopropyl)methyl)cyclopropyl)octanoic acid | 28399-31-7 | MFCD09265251 | 98.0% | 308.50 g/mol | C20H36O2 | 5 MG
SDP
Catalog No. 5000263823
Encompass_Preferred
DCP-LA (FR236924) is a linoleic acid-derived small molecule activator of PKCε used in biochemical and cell-signaling research. It is a lipophilic carboxylic acid (C20H36O2, MW 308.50) typically supplied with high purity and available as solid quantities or as a DMSO solution. Store under cold conditions to preserve stability and follow laboratory safety protocols for handling bioactive lipids.
  • Selective activator of PKCε for signaling and apoptosis studies.
  • High purity (≥98.0%) suitable for research applications.
  • Available in multiple package formats including mg quantities and DMSO solutions.
  • Molecular formula C20H36O2; molecular weight 308.50 g/mol.
  • Store cold; solutions require deep-freeze storage for extended stability.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-63823 Supplier Medchemexpress LLC Supplier No. HY1085995MG
DCP-LA (FR236924) is a linoleic acid-derived small molecule activator of PKCε used in biochemical and cell-signaling research. It is a lipophilic carboxylic acid (C20H36O2, MW 308.50) typically supplied with high purity and available as solid quantities or as a DMSO solution. Store under cold conditions to preserve stability and follow laboratory safety protocols for handling bioactive lipids.
  • Selective activator of PKCε for signaling and apoptosis studies.
  • High purity (≥98.0%) suitable for research applications.
  • Available in multiple package formats including mg quantities and DMSO solutions.
  • Molecular formula C20H36O2; molecular weight 308.50 g/mol.
  • Store cold; solutions require deep-freeze storage for extended stability.
  • For research use only; not for human or clinical use.