Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-[2-(2-Pyridyl)phenyl]-5H-pyrido[3,2-b]indole 98.0+%, TCI America™
SDP

Catalog No. P2307100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2307100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2307100MG Supplier TCI America Supplier No. P2307100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1251503-35-1
Molecular Formula C22H15N3
Molecular Weight (g/mol) 321.383
InChI Key ROQWGQBJRGUPAG-UHFFFAOYSA-N
PubChem CID 51051482
IUPAC Name 8-(2-pyridin-2-ylphenyl)-5H-pyrido[3,2-b]indole
SMILES C1=CC=C(C(=C1)C2=CC3=C(C=C2)NC4=C3N=CC=C4)C5=CC=CC=N5

Specifications

Quantity 100 mg
Formula Weight 321.38
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 8-[2-(2-Pyridyl)phenyl]-5H-pyrido[3,2-b]indole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.