Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-(2,4-dichlorophenyl)-N-(1,3-dimethoxypropan-2-yl)-2,7-dimethylpyrazolo[5,1-f]triazin-4-amine | 202578-52-7 | 98.7% | 100 MG
SDP

Catalog No. 5000232544
Encompass_Preferred

DMP 696 is a selective corticotropin-releasing hormone receptor 1 (CRHR1) antagonist used in research on anxiety and depression. Supplied as a solid research-grade compound, it is characterized by a molecular formula C18H21Cl2N5O2 and a molecular weight of 410.30 g/mol. Store and handle according to supplier recommendations for research use only.

  • Selective CRHR1 antagonist for anxiety and depression research.
  • High reported purity (≈98.7% based on supplier documentation).
  • Solid form with multiple pack sizes available, including 100 MG.
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation recommended.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).

Catalog No. 50-002-32544 Supplier Medchemexpress LLC Supplier No. HY12131100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DMP 696 is a selective corticotropin-releasing hormone receptor 1 (CRHR1) antagonist used in research on anxiety and depression. Supplied as a solid research-grade compound, it is characterized by a molecular formula C18H21Cl2N5O2 and a molecular weight of 410.30 g/mol. Store and handle according to supplier recommendations for research use only.

  • Selective CRHR1 antagonist for anxiety and depression research.
  • High reported purity (≈98.7% based on supplier documentation).
  • Solid form with multiple pack sizes available, including 100 MG.
  • Soluble in DMSO at 100 mg/mL; ultrasonic agitation recommended.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.