Learn More
Chemglass Life Sciences 5.82" ID x 8.20" OD x 1.20" Height White PTFE Support Ring, 70° "V" Cut
SDP
Catalog No. 50997780
Encompass_Preferred
5.82" ID x 8.20" OD x 1.20" Height White PTFE Support Ring, 70¿ "V" Cut. PTFE support ring used to support round bottom flasks or cylindrical reaction vessels.PLEASE NOTE: PTFE SUPPORT RINGS CAN BE CUT. PLEASE CALL FOR PRICING
Catalog No. 50-997-780 Supplier Chemglass Life Sciences Supplier No. CG305510
5.82" ID x 8.20" OD x 1.20" Height White PTFE Support Ring, 70¿ "V" Cut. PTFE support ring used to support round bottom flasks or cylindrical reaction vessels.PLEASE NOTE: PTFE SUPPORT RINGS CAN BE CUT. PLEASE CALL FOR PRICING