Learn More
Chemglass Life Sciences 5.82" ID x 8.20" OD x 1.20" Height White PTFE Support Ring
SDP
Catalog No. 50872572
Encompass_Preferred
5.82" ID x 8.20" OD x 1.20" Height White PTFE Support Ring. PTFE support ring used to support round bottom flasks or cylindrical reaction vessels.PLEASE NOTE: PTFE SUPPORT RINGS CAN BE CUT. PLEASE CALL FOR PRICING
Catalog No. 50-872-572 Supplier Chemglass Life Sciences Supplier No. CG305508
5.82" ID x 8.20" OD x 1.20" Height White PTFE Support Ring. PTFE support ring used to support round bottom flasks or cylindrical reaction vessels.PLEASE NOTE: PTFE SUPPORT RINGS CAN BE CUT. PLEASE CALL FOR PRICING