Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 8-[3-(4-fluorophenoxy)propyl]-1-phenyl-1,3,8-triazaspiro[4,5]decan-4-one | 510-74-7 | 99.2% | 383.46 | C22H26FN3O2 | 25 MG
SDP

Catalog No. 5000211675
Encompass_Preferred

Spiramide (AMI-193) is a research-grade small molecule antagonist of serotonin 5-HT2 and dopamine D2 receptors. It is supplied as a solid or as a DMSO solution for in vitro pharmacology, receptor binding, and functional assays. The compound demonstrates high potency and selectivity and is intended for laboratory research use only.

  • Potent antagonist of 5-HT2 and dopamine D2 receptors (Ki = 2 nM and 3 nM).
  • High selectivity versus 5-HT1C receptors (Ki = 4,300 nM).
  • Available as solid and as 10 mM solution in DMSO in multiple small pack sizes.
  • Purity typically greater than 99% by supplier analysis.
  • Molecular weight 383.46; CAS 510-74-7.
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-11675 Supplier Medchemexpress LLC Supplier No. HY10097125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Spiramide (AMI-193) is a research-grade small molecule antagonist of serotonin 5-HT2 and dopamine D2 receptors. It is supplied as a solid or as a DMSO solution for in vitro pharmacology, receptor binding, and functional assays. The compound demonstrates high potency and selectivity and is intended for laboratory research use only.

  • Potent antagonist of 5-HT2 and dopamine D2 receptors (Ki = 2 nM and 3 nM).
  • High selectivity versus 5-HT1C receptors (Ki = 4,300 nM).
  • Available as solid and as 10 mM solution in DMSO in multiple small pack sizes.
  • Purity typically greater than 99% by supplier analysis.
  • Molecular weight 383.46; CAS 510-74-7.
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.