Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-(3-Chlorostyryl)caffeine, Tocris Bioscience™

Catalog No. 330650
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-065-0 50 mg
33-061-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-065-0 Supplier R&D Systems Supplier No. 3306/50

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Selective A2A antagonist. Also MAO-B inhibitor

Selective adenosine A2A receptor antagonist and monoamine oxidase B (MAO-B) inhibitor (Ki values are 54 and 28200 nM at rat A2A and A1 receptors respectively and Ki ∽ 100 nM at MAO-B). Potently protects against quinolinic acid -induced (Cat. No. 0225) neuronal damage and is neuroprotective in the MPTP model of Parkinson's disease.

Chemical Identifiers

CAS 147700-11-6
Molecular Formula C32H30Cl2N8O4
Molecular Weight (g/mol) 661.544
InChI Key YMUNFKYGBSLPLQ-UHFFFAOYSA-N
Synonym 1h-purine-2,6-dione,8-1e-2-3-chlorophenyl ethenyl-3,7-dihydro-1,3,7-trimethyl, acmc-20n59d
PubChem CID 57348597
IUPAC Name 8-[2-(3-chlorophenyl)ethenyl]-1,3,7-trimethylpurine-2,6-dione
SMILES CN1C(=NC2=C1C(=O)N(C(=O)N2C)C)C=CC3=CC(=CC=C3)Cl.CN1C(=NC2=C1C(=O)N(C(=O)N2C)C)C=CC3=CC(=CC=C3)Cl

Specifications

Quantity 50 mg
Formula Weight 330.77
Percent Purity >99%
Chemical Name or Material 8-(3-Chlorostyryl)caffeine
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.