Learn More
Cayman Chemical 8- 3-ChlorostyrylcaffeIn 50mg
SDP
Catalog No. 502989128
Encompass_Preferred
An adenosine A2 receptor antagonist (Ki = 54 nM); selective for adenosine A2 over A1 receptors (Ki = 28 µM); reverses APEC-induced locomotor depression in mice (ED50 = 16 UG/kg, i.p.); increases locomotion in mice at 5 mg/kg; reverses L-DOPA-induced shortening of the rotational motor response and increases in striatal adenosine A2 receptor levels in a rat model of 6-OHDA-induced Parkinson's disease at 5 mg/kg; inhibits MAO-B (Ki = 100 nM); reduces MAO-B activity in brain mitochondrial preparations from wild-type and A2A-/- mice
Catalog No. 50-298-9128 Supplier Cayman Chemical Supplier No. 2970450 MG
An adenosine A2 receptor antagonist (Ki = 54 nM); selective for adenosine A2 over A1 receptors (Ki = 28 µM); reverses APEC-induced locomotor depression in mice (ED50 = 16 UG/kg, i.p.); increases locomotion in mice at 5 mg/kg; reverses L-DOPA-induced shortening of the rotational motor response and increases in striatal adenosine A2 receptor levels in a rat model of 6-OHDA-induced Parkinson's disease at 5 mg/kg; inhibits MAO-B (Ki = 100 nM); reduces MAO-B activity in brain mitochondrial preparations from wild-type and A2A-/- mice