Learn More
Medchemexpress LLC 8-[4-(1-aminocyclobutyl)phenyl]-9-phenyl-2H-triazolo[3,4-f]naphthyridin-3-one | 1032349-77-1 | 99.7% | 443.93 | C25H22ClN5O | 1 ML
SDP
Catalog No. 5000241929
Encompass_Preferred
MK-2206 is an orally active, potent, and selective allosteric inhibitor of Akt provided as a 10 mM solution (1 mL) for research applications. It is used to probe Akt signaling and downstream processes such as apoptosis and autophagy in cell-based and organoid models.
  • Highly potent and selective allosteric Akt inhibition suitable for signaling studies.
  • Ready-to-use 10 mM solution (1 mL) for convenient dosing and assay setup.
  • High purity (>98%) for reliable experimental results.
  • Applicable to studies of apoptosis, autophagy, and organoid biology.
  • Documented chemical identity and molecular data for traceability.
Catalog No. 50-002-41929 Supplier Medchemexpress LLC Supplier No. HY10823210MM 1ML
MK-2206 is an orally active, potent, and selective allosteric inhibitor of Akt provided as a 10 mM solution (1 mL) for research applications. It is used to probe Akt signaling and downstream processes such as apoptosis and autophagy in cell-based and organoid models.
  • Highly potent and selective allosteric Akt inhibition suitable for signaling studies.
  • Ready-to-use 10 mM solution (1 mL) for convenient dosing and assay setup.
  • High purity (>98%) for reliable experimental results.
  • Applicable to studies of apoptosis, autophagy, and organoid biology.
  • Documented chemical identity and molecular data for traceability.