Learn More
Medchemexpress LLC 8-(4-chlorophenyl)-6-(2-methyl-2H-indazol-5-yl)-2-[(2,2,2-trifluoroethyl)amino]pyrido[4,3-d]pyrimidin-7(6H)-one | 2377491-54-6 | 99.5% | 484.86 | C23H16ClF3N6O | 5 MG
SDP
Catalog No. 5000221684
Encompass_Preferred
AGI-43192 is a potent small-molecule inhibitor of methionine adenosyltransferase 2A (MAT2A). It is orally active with limited blood-brain barrier penetration and is supplied as a light yellow solid for research use in biochemical and pharmacological studies.
  • Potent MAT2A inhibition shown in biochemical assays.
  • Orally active pharmacokinetic profile suitable for in vivo studies.
  • Limited blood-brain barrier penetration for peripheral target engagement.
  • High purity for reliable experimental results.
  • Soluble in DMSO and compatible with published in vivo formulations.
  • Stable under recommended storage conditions (powder -20°C).
Catalog No. 50-002-21684 Supplier Medchemexpress LLC Supplier No. HY1457775MG
AGI-43192 is a potent small-molecule inhibitor of methionine adenosyltransferase 2A (MAT2A). It is orally active with limited blood-brain barrier penetration and is supplied as a light yellow solid for research use in biochemical and pharmacological studies.
  • Potent MAT2A inhibition shown in biochemical assays.
  • Orally active pharmacokinetic profile suitable for in vivo studies.
  • Limited blood-brain barrier penetration for peripheral target engagement.
  • High purity for reliable experimental results.
  • Soluble in DMSO and compatible with published in vivo formulations.
  • Stable under recommended storage conditions (powder -20°C).