Learn More
Medchemexpress LLC 8-(4-chlorophenylthio)adenosine 3',5'-cyclic monophosphate monosodium salt | 93882-12-3 | 99.9% | 493.79 g/mol | C16H14ClN5NaO6PS | 5 MG
SDP
Catalog No. 5000264162
Encompass_Preferred
8-CPT-Cyclic AMP sodium is a membrane-permeable cyclic AMP analog that selectively activates cAMP-dependent protein kinase (PKA) and inhibits certain phosphodiesterases. Supplied as the monosodium salt in solid form, it is used to elevate intracellular cAMP levels and to probe cAMP/PKA signaling pathways in biochemical and cell-based assays.
  • Selective activator of cAMP-dependent protein kinase.
  • Membrane-permeable cyclic AMP analog (monosodium salt).
  • High purity (~99.9%) for reproducible experimental results.
  • Soluble in DMSO and water with sonication and warming as needed.
  • Solid storage at -20°C; in solution store at -80°C for long-term stability.
  • Available in small-scale solid quantities and in ready-to-use solution format.
Catalog No. 50-002-64162 Supplier Medchemexpress LLC Supplier No. HY1116735MG
8-CPT-Cyclic AMP sodium is a membrane-permeable cyclic AMP analog that selectively activates cAMP-dependent protein kinase (PKA) and inhibits certain phosphodiesterases. Supplied as the monosodium salt in solid form, it is used to elevate intracellular cAMP levels and to probe cAMP/PKA signaling pathways in biochemical and cell-based assays.
  • Selective activator of cAMP-dependent protein kinase.
  • Membrane-permeable cyclic AMP analog (monosodium salt).
  • High purity (~99.9%) for reproducible experimental results.
  • Soluble in DMSO and water with sonication and warming as needed.
  • Solid storage at -20°C; in solution store at -80°C for long-term stability.
  • Available in small-scale solid quantities and in ready-to-use solution format.