Learn More
Medchemexpress LLC 8-(4-iodophenyl)-1,3,5,7-tetramethyl BODIPY | 250734-47-5 | 98.5% | 450.07 g·mol⁻¹ | C19H18BF2IN2 | 1 G
SDP
Catalog No. 5000281293
Encompass_Preferred
8-(4-iodophenyl)-1,3,5,7-tetramethyl BODIPY is an iodinated BODIPY-family fluorescent dye used in research applications as a probe for GFP-labeled muscarinic M1 receptors and for general fluorescent imaging and spectroscopy. It is provided as a high-purity reagent appropriate for laboratory assays, labeling, and microscopy work.
  • Iodinated BODIPY fluorophore for fluorescent labeling and imaging.
  • Suitable for probing GFP-labeled muscarinic M1 receptors in biological assays.
  • High chemical purity (~98%) for reproducible experimental results.
  • Molecular weight 450.07 g·mol⁻¹; formula C19H18BF2IN2.
  • Typically supplied as a colored solid and stored protected from light at cool temperatures.
Catalog No. 50-002-81293 Supplier Medchemexpress LLC Supplier No. HYW2481191G
8-(4-iodophenyl)-1,3,5,7-tetramethyl BODIPY is an iodinated BODIPY-family fluorescent dye used in research applications as a probe for GFP-labeled muscarinic M1 receptors and for general fluorescent imaging and spectroscopy. It is provided as a high-purity reagent appropriate for laboratory assays, labeling, and microscopy work.
  • Iodinated BODIPY fluorophore for fluorescent labeling and imaging.
  • Suitable for probing GFP-labeled muscarinic M1 receptors in biological assays.
  • High chemical purity (~98%) for reproducible experimental results.
  • Molecular weight 450.07 g·mol⁻¹; formula C19H18BF2IN2.
  • Typically supplied as a colored solid and stored protected from light at cool temperatures.