Learn More
Medchemexpress LLC 8-(4-iodophenyl)-1,3,5,7-tetramethyl BODIPY | 250734-47-5 | MFCD29905036 | >98.0% | 450.07 g/mol | C19H18BF2IN2 | 50 MG
SDP
Catalog No. 5000280696
Encompass_Preferred
8-(4-iodophenyl)-1,3,5,7-tetramethyl BODIPY is an iodinated BODIPY-family fluorescent dye used as a probe in fluorescence imaging. It has been reported as a fluorescent probe for GFP-labeled muscarinic M1 receptor. The compound is a yellow-orange solid with formula C19H18BF2IN2, molecular weight ~450.07 g/mol, and is typically supplied with high purity for research applications.
  • Iodinated BODIPY core provides strong fluorescence suitable for imaging.
  • Reported application as a probe for GFP-labeled muscarinic M1 receptor.
  • High purity >98.0% (HPLC).
  • Yellow to orange solid appearance.
  • Molecular weight 450.07 g/mol; formula C19H18BF2IN2.
  • Store protected from light; in solvent -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-80696 Supplier Medchemexpress LLC Supplier No. HYW24811950MG
8-(4-iodophenyl)-1,3,5,7-tetramethyl BODIPY is an iodinated BODIPY-family fluorescent dye used as a probe in fluorescence imaging. It has been reported as a fluorescent probe for GFP-labeled muscarinic M1 receptor. The compound is a yellow-orange solid with formula C19H18BF2IN2, molecular weight ~450.07 g/mol, and is typically supplied with high purity for research applications.
  • Iodinated BODIPY core provides strong fluorescence suitable for imaging.
  • Reported application as a probe for GFP-labeled muscarinic M1 receptor.
  • High purity >98.0% (HPLC).
  • Yellow to orange solid appearance.
  • Molecular weight 450.07 g/mol; formula C19H18BF2IN2.
  • Store protected from light; in solvent -80°C (6 months) or -20°C (1 month).