Learn More
Medchemexpress LLC 8-(4-iodophenyl)-1,3,5,7-tetramethyl BODIPY | 250734-47-5 | MFCD29905036 | 98.0% | 450.07 g/mol | C19H18BF2IN2 | 500 MG
SDP
Catalog No. 5000281351
Encompass_Preferred
8-(4-iodophenyl)-1,3,5,7-tetramethyl BODIPY is an iodinated BODIPY fluorescent dye used as a probe for GFP-labeled muscarinic M1 receptors in fluorescence microscopy and receptor-binding assays. It is supplied as a solid research reagent with reported high purity for imaging and analytical workflows.
  • Iodinated BODIPY fluorophore optimized for GFP-labeled muscarinic M1 receptor studies.
  • High purity suitable for imaging and analytical applications.
  • Solid form for convenient storage and handling.
  • Compatible with fluorescence microscopy and receptor-binding assays.
  • Available in 500 MG quantity for preparative or multiple-sample use.
Catalog No. 50-002-81351 Supplier Medchemexpress LLC Supplier No. HYW248119500MG
8-(4-iodophenyl)-1,3,5,7-tetramethyl BODIPY is an iodinated BODIPY fluorescent dye used as a probe for GFP-labeled muscarinic M1 receptors in fluorescence microscopy and receptor-binding assays. It is supplied as a solid research reagent with reported high purity for imaging and analytical workflows.
  • Iodinated BODIPY fluorophore optimized for GFP-labeled muscarinic M1 receptor studies.
  • High purity suitable for imaging and analytical applications.
  • Solid form for convenient storage and handling.
  • Compatible with fluorescence microscopy and receptor-binding assays.
  • Available in 500 MG quantity for preparative or multiple-sample use.